Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCGCCTGGTTTTCGAGCCTTA[C/T]TGATTAGAGTTTGGACCCCTTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FLYWCH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLYWCH1 - FLYWCH-type zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308068.1 | Intron | NP_001294997.1 | ||||
NM_020912.1 | Intron | NP_065963.1 | ||||
NM_032296.2 | Intron | NP_115672.2 | ||||
XM_006720962.3 | Intron | XP_006721025.1 | ||||
XM_011522701.2 | Intron | XP_011521003.1 | ||||
XM_011522702.2 | Intron | XP_011521004.1 | ||||
XM_011522703.2 | Intron | XP_011521005.1 | ||||
XM_011522704.2 | Intron | XP_011521006.1 | ||||
XM_011522705.2 | Intron | XP_011521007.1 | ||||
XM_011522707.2 | Intron | XP_011521009.1 | ||||
XM_017023772.1 | Intron | XP_016879261.1 |