Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTACAACTTCCGACTTGCATC[A/C]CTGCGGTGGCGCTAAACAGCGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC5 - armadillo repeat containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105247.1 | 413 | UTR 5 | NP_001098717.1 | |||
NM_001288767.1 | 413 | Intron | NP_001275696.1 | |||
NM_001301820.1 | 413 | Intron | NP_001288749.1 | |||
NM_024742.2 | 413 | UTR 5 | NP_079018.1 | |||
XM_006721091.2 | 413 | Intron | XP_006721154.1 |