Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCGCTTCATCACTTCTCGAAAA[C/T]GCTCGCCCTAGAATGGCAAAGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929989 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929989 - lipopolysaccharide-induced tumor necrosis factor-alpha factor-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP7 - ubiquitin specific peptidase 7 (herpes virus-associated) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286457.1 | 3246 | Missense Mutation | NP_001273386.1 | |||
NM_001286458.1 | 3246 | Missense Mutation | NP_001273387.1 | |||
NM_001321858.1 | 3246 | Missense Mutation | NP_001308787.1 | |||
NM_003470.2 | 3246 | Missense Mutation | NP_003461.2 | |||
XM_017023652.1 | 3246 | Missense Mutation | XP_016879141.1 | |||
XM_017023653.1 | 3246 | Missense Mutation | XP_016879142.1 |