Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTTTCCCCCATGGTGATGTGG[A/G]TGCTCTGGTTGGGGCAGAGCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRG1 - adhesion G protein-coupled receptor G1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ADGRG3 - adhesion G protein-coupled receptor G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308360.1 | Intron | NP_001295289.1 | ||||
NM_170776.4 | Intron | NP_740746.4 | ||||
XM_005255842.3 | Intron | XP_005255899.1 | ||||
XM_006721170.3 | Intron | XP_006721233.1 | ||||
XM_011522951.2 | Intron | XP_011521253.1 | ||||
XM_011522953.2 | Intron | XP_011521255.1 | ||||
XM_011522954.1 | Intron | XP_011521256.1 |