Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTACTTTTAGGTAAGCATACAT[G/T]AAAGTTTTCGAGATATTATCTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603008 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH8 - cadherin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001796.4 | 5367 | Intron | NP_001787.2 | |||
XM_005255760.3 | 5367 | UTR 3 | XP_005255817.1 | |||
XM_017022829.1 | 5367 | Intron | XP_016878318.1 |