Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACTGAATCCAAACTCTTAAAT[A/G]AGATGGCGATCAAAGAGCGCGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616074 MIM: 607884 | ||||||||||||||||||||
Literature Links: |
CKLF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CKLF - chemokine-like factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040138.2 | 386 | Intron | NP_001035228.1 | |||
NM_016326.3 | 386 | Intron | NP_057410.1 | |||
NM_016951.3 | 386 | Intron | NP_058647.1 | |||
NM_181640.2 | 386 | Intron | NP_857591.1 | |||
NM_181641.2 | 386 | Intron | NP_857592.1 |
CKLF-CMTM1 - CKLF-CMTM1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202509.1 | 386 | Intron | NP_001189438.1 | |||
NM_001204098.1 | 386 | Intron | NP_001191027.1 | |||
NM_001204099.1 | 386 | Intron | NP_001191028.1 |
CMTM1 - CKLF like MARVEL transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052999.3 | 386 | Missense Mutation | AAG,GAG | K,E 107 | NP_443725.3 | |
NM_181268.2 | 386 | Missense Mutation | AAG,GAG | K,E 107 | NP_851785.2 | |
NM_181269.2 | 386 | Intron | NP_851786.1 | |||
NM_181270.2 | 386 | Intron | NP_851787.1 | |||
NM_181271.2 | 386 | Intron | NP_851788.1 | |||
NM_181272.2 | 386 | Intron | NP_851789.1 | |||
NM_181283.2 | 386 | Intron | NP_851800.1 | |||
NM_181296.2 | 386 | Intron | NP_851813.1 |