Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATGCCTAGCAGGCTGTTCTTT[G/T]ACAACTTGGGTGCCATCCTCACCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606881 MIM: 600477 | ||||||||||||||||||||
Literature Links: |
FHOD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FHOD1 - formin homology 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A5 - solute carrier family 9 member A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323971.1 | 1854 | Missense Mutation | GAC,TAC | D,Y 130 | NP_001310900.1 | |
NM_001323972.1 | 1854 | Intron | NP_001310901.1 | |||
NM_001323973.1 | 1854 | Missense Mutation | GAC,TAC | D,Y 130 | NP_001310902.1 | |
NM_001323974.1 | 1854 | UTR 5 | NP_001310903.1 | |||
NM_001323975.1 | 1854 | UTR 5 | NP_001310904.1 | |||
NM_004594.2 | 1854 | Missense Mutation | GAC,TAC | D,Y 130 | NP_004585.1 | |
XM_017023594.1 | 1854 | Missense Mutation | GAC,TAC | D,Y 130 | XP_016879083.1 |