Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGGAGAAGACGATGTCCACATC[A/G]GTCCCTGTCACGGACTTTCCGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616957 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC36 - leucine rich repeat containing 36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPPP3 - tubulin polymerization promoting protein family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015964.3 | 321 | Silent Mutation | ACC,ACT | T,T 53 | NP_057048.2 | |
NM_016140.3 | 321 | Silent Mutation | ACC,ACT | T,T 53 | NP_057224.2 |
ZDHHC1 - zinc finger DHHC-type containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |