Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTTCAGGCAACCTTTTCATCA[C/T]TTCTCATCTCTTCTTTCAATGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604708 MIM: 125860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFAT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFAT5 - nuclear factor of activated T-cells 5, tonicity-responsive | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NQO1 - NAD(P)H quinone dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000903.2 | 2231 | UTR 3 | NP_000894.1 | |||
NM_001025433.1 | 2231 | UTR 3 | NP_001020604.1 | |||
NM_001025434.1 | 2231 | UTR 3 | NP_001020605.1 | |||
NM_001286137.1 | 2231 | UTR 3 | NP_001273066.1 |