Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAATGTACTAATGCAAGGACTG[G/A]TAAGACTTAAGATTCACATAACGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFWD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFWD3 - ring finger and WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018124.3 | 4302 | UTR 3 | NP_060594.3 | |||
XM_005256021.4 | 4302 | UTR 3 | XP_005256078.1 | |||
XM_005256022.4 | 4302 | UTR 3 | XP_005256079.1 | |||
XM_006721228.3 | 4302 | UTR 3 | XP_006721291.1 | |||
XM_011523191.2 | 4302 | UTR 3 | XP_011521493.1 | |||
XM_017023391.1 | 4302 | UTR 3 | XP_016878880.1 | |||
XM_017023392.1 | 4302 | UTR 3 | XP_016878881.1 |