Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCCAGAGTCTCCTAGATTTATT[G/T]AATAAAATAATCTGGAAGAAATAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM170A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM170A - transmembrane protein 170A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304996.1 | Intron | NP_001291925.1 | ||||
NM_001304997.1 | Intron | NP_001291926.1 | ||||
NM_001304998.1 | Intron | NP_001291927.1 | ||||
NM_145254.2 | Intron | NP_660297.1 | ||||
XM_011522869.2 | Intron | XP_011521171.1 | ||||
XM_017022939.1 | Intron | XP_016878428.1 | ||||
XM_017022940.1 | Intron | XP_016878429.1 | ||||
XM_017022941.1 | Intron | XP_016878430.1 | ||||
XM_017022942.1 | Intron | XP_016878431.1 | ||||
XM_017022943.1 | Intron | XP_016878432.1 |