Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATCCTCCCTGCTCACAAAACA[A/G]CCAGCTACTTGGTTTTCTAAAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 177075 MIM: 605131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371352 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371352 - uncharacterized LOC105371352 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAF - MAF bZIP transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031804.2 | Intron | NP_001026974.1 | ||||
NM_005360.4 | Intron | NP_005351.2 | ||||
XM_017023233.1 | Intron | XP_016878722.1 | ||||
XM_017023234.1 | Intron | XP_016878723.1 | ||||
XM_017023235.1 | Intron | XP_016878724.1 |
WWOX - WW domain containing oxidoreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291997.1 | Intron | NP_001278926.1 | ||||
NM_016373.3 | Intron | NP_057457.1 | ||||
NM_130791.3 | Intron | NP_570607.1 | ||||
XM_011523101.2 | Intron | XP_011521403.1 | ||||
XM_011523103.2 | Intron | XP_011521405.1 | ||||
XM_011523104.2 | Intron | XP_011521406.1 | ||||
XM_011523105.2 | Intron | XP_011521407.1 | ||||
XM_017023278.1 | Intron | XP_016878767.1 | ||||
XM_017023279.1 | Intron | XP_016878768.1 |