Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGCCGGGAGCCTGACCCACTT[A/C]CCTCTTCCCTCCCGGGACCCACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNLRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYNLRB2 - dynein light chain roadblock-type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305017.1 | 74 | Missense Mutation | ACC,CCC | T,P 15 | NP_001291946.1 | |
NM_130897.2 | 74 | Intron | NP_570967.1 | |||
XM_006721295.3 | 74 | Intron | XP_006721358.1 | |||
XM_006721296.3 | 74 | Intron | XP_006721359.1 | |||
XM_011523370.2 | 74 | Intron | XP_011521672.1 |
LOC102724084 - uncharacterized LOC102724084 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |