Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATTTCCTTTCCAAACACAGATC[C/T]GGGACACACGTTACAAGCGTACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607894 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKD1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKD1L2 - polycystin 1 like 2 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076780.1 | Intron | NP_001070248.1 | ||||
NM_001278423.1 | Intron | NP_001265352.1 | ||||
NM_001278425.1 | Intron | NP_001265354.1 | ||||
NM_052892.3 | Intron | NP_443124.3 |