Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGACCCTCAGCACGCCGCGCGT[C/T]TGCTGCGAGCGCTCAGCTCTTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605379 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAN - gigaxonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022041.3 | 194 | Silent Mutation | CTG,TTG | L,L 16 | NP_071324.1 | |
XM_017023734.1 | 194 | UTR 5 | XP_016879223.1 |