Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGCTGCAGGAGGCAGATCATC[C/T]GGGCTCTGTGCCTCTTGCTACTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605322 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376777 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105376777 - uncharacterized LOC105376777 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011523512.2 | 315 | Intron | XP_011521814.1 |
WFDC1 - WAP four-disulfide core domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282466.1 | 315 | Missense Mutation | CGG,TGG | R,W 17 | NP_001269395.1 | |
NM_001282467.1 | 315 | Missense Mutation | CGG,TGG | R,W 17 | NP_001269396.1 | |
NM_021197.3 | 315 | Missense Mutation | CGG,TGG | R,W 17 | NP_067020.2 | |
XM_005256079.3 | 315 | Missense Mutation | CGG,TGG | R,W 17 | XP_005256136.1 |