Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACAAGTCTCAAGTGTTTGTTGC[C/G]CAACAGGCCGGGTTTTCTGTGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611675 | ||||||||||||||||||||
Literature Links: |
FAM92B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM92B - family with sequence similarity 92 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198491.2 | 1498 | UTR 3 | NP_940893.1 | |||
XM_005255931.2 | 1498 | Intron | XP_005255988.1 | |||
XM_011523063.1 | 1498 | Intron | XP_011521365.1 | |||
XM_017023198.1 | 1498 | Intron | XP_016878687.1 |
KIAA0513 - KIAA0513 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |