Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATACGTACACAGAGGGCACATA[C/T]CCCCACACCCATGCATGCAGGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609759 MIM: 615805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM7 - PR domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098173.1 | 2581 | UTR 3 | NP_001091643.1 | |||
XM_005256274.3 | 2581 | UTR 3 | XP_005256331.1 | |||
XM_011522829.1 | 2581 | UTR 3 | XP_011521131.1 | |||
XM_011522831.2 | 2581 | Intron | XP_011521133.1 | |||
XM_017022882.1 | 2581 | Intron | XP_016878371.1 | |||
XM_017022883.1 | 2581 | Intron | XP_016878372.1 | |||
XM_017022884.1 | 2581 | Intron | XP_016878373.1 |
URAHP - urate (hydroxyiso-) hydrolase, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |