Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGCCAGGCCAGCTCCCCCTCTG[C/T]CAGGTGCATTCTGTCTGGTGACTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606236 MIM: 602050 MIM: 615128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPSCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASPSCR1 - ASPSCR1, UBX domain containing tether for SLC2A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC45 - leucine rich repeat containing 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144999.3 | Intron | NP_659436.1 |
RAC3 - ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRA13 - stimulated by retinoic acid 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |