Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAACACCTGAGCCTGATGGCAAG[A/G]ACCCATGAAACTTTAGTGCCAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603644 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADPRM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADPRM - ADP-ribose/CDP-alcohol diphosphatase, manganese dependent | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020233.4 | Intron | NP_064618.3 | ||||
XM_005256738.2 | Intron | XP_005256795.1 |
SCO1 - SCO1 cytochrome c oxidase assembly protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |