Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAATTACGCGTTTTTATGTATT[C/T]GGTGTTTCAGGTTCAGTCATATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIGL - phosphatidylinositol glycan anchor biosynthesis class L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004278.3 | Intron | NP_004269.1 | ||||
XM_011524080.2 | Intron | XP_011522382.1 | ||||
XM_017025349.1 | Intron | XP_016880838.1 | ||||
XM_017025350.1 | Intron | XP_016880839.1 | ||||
XM_017025351.1 | Intron | XP_016880840.1 | ||||
XM_017025352.1 | Intron | XP_016880841.1 | ||||
XM_017025353.1 | Intron | XP_016880842.1 | ||||
XM_017025354.1 | Intron | XP_016880843.1 | ||||
XM_017025355.1 | Intron | XP_016880844.1 | ||||
XM_017025356.1 | Intron | XP_016880845.1 |