Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACATATATTACATATATAGGAT[C/T]TCTCTCCAGTGTGAGTGGTTTGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF287 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF287 - zinc finger protein 287 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020653.2 | 2222 | Missense Mutation | AAA,AGA | K,R 590 | NP_065704.2 | |
XM_011523968.2 | 2222 | Missense Mutation | AAA,AGA | K,R 590 | XP_011522270.1 | |
XM_017024887.1 | 2222 | Missense Mutation | AAA,AGA | K,R 590 | XP_016880376.1 |