Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAAGAATTGATATGTGAGAAAA[G/A]CCAATGAATTTTAGAGCAATCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610963 MIM: 606477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMG6 - SMG6, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRR - serine racemase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304803.1 | Intron | NP_001291732.1 | ||||
NM_021947.2 | Intron | NP_068766.1 | ||||
XM_006721565.3 | Intron | XP_006721628.1 | ||||
XM_006721566.3 | Intron | XP_006721629.1 | ||||
XM_011523974.2 | Intron | XP_011522276.1 |