Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATTCCACTTAAACAGCCAAAT[A/T]GACTTAAAAGGGCAACATAGATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ULK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ULK2 - unc-51 like autophagy activating kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142610.1 | 8623 | Intron | NP_001136082.1 | |||
NM_014683.3 | 8623 | UTR 3 | NP_055498.3 | |||
XM_011524087.2 | 8623 | Intron | XP_011522389.1 | |||
XM_017025424.1 | 8623 | Intron | XP_016880913.1 | |||
XM_017025425.1 | 8623 | Intron | XP_016880914.1 | |||
XM_017025426.1 | 8623 | Intron | XP_016880915.1 | |||
XM_017025427.1 | 8623 | Intron | XP_016880916.1 | |||
XM_017025428.1 | 8623 | Intron | XP_016880917.1 |