Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCGTGTGTTCAGGAGACCTCAG[A/G]GGTCCGTGTAGACTGTCCTCTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP1GAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAP1GAP2 - RAP1 GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100398.1 | Intron | NP_001093868.1 | ||||
NM_015085.4 | Intron | NP_055900.4 | ||||
XM_005256542.2 | Intron | XP_005256599.1 | ||||
XM_006721477.3 | Intron | XP_006721540.1 | ||||
XM_011523738.1 | Intron | XP_011522040.1 | ||||
XM_011523739.1 | Intron | XP_011522041.1 | ||||
XM_011523740.2 | Intron | XP_011522042.1 | ||||
XM_011523741.2 | Intron | XP_011522043.1 | ||||
XM_011523742.2 | Intron | XP_011522044.1 | ||||
XM_011523743.2 | Intron | XP_011522045.1 | ||||
XM_011523744.2 | Intron | XP_011522046.1 | ||||
XM_011523745.2 | Intron | XP_011522047.1 | ||||
XM_017024370.1 | Intron | XP_016879859.1 | ||||
XM_017024371.1 | Intron | XP_016879860.1 |