Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTTACGACTGGGAGTGTGGCTT[G/T]ATTCTTGCAGTTGGCTTCACTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611627 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM57A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM57A - family with sequence similarity 57 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318006.1 | Intron | NP_001304935.1 | ||||
NM_001318007.1 | Intron | NP_001304936.1 | ||||
NM_001318008.1 | Intron | NP_001304937.1 | ||||
NM_024792.2 | Intron | NP_079068.1 |