Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTCTGAGGTACACACCGGAGAC[C/T]CCACTCTGTTCCCAGCGCTCACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601929 MIM: 600845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP2A3 - ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005173.3 | 3338 | UTR 3 | NP_005164.2 | |||
NM_174953.2 | 3338 | UTR 3 | NP_777613.1 | |||
NM_174954.2 | 3338 | UTR 3 | NP_777614.1 | |||
NM_174955.2 | 3338 | Missense Mutation | GAG,GGG | E,G 1036 | NP_777615.1 | |
NM_174956.2 | 3338 | UTR 3 | NP_777616.1 | |||
NM_174957.2 | 3338 | UTR 3 | NP_777617.1 | |||
NM_174958.2 | 3338 | UTR 3 | NP_777618.1 | |||
XM_011523881.2 | 3338 | Intron | XP_011522183.1 | |||
XM_011523882.2 | 3338 | Intron | XP_011522184.1 | |||
XM_011523884.2 | 3338 | Intron | XP_011522186.1 | |||
XM_011523885.1 | 3338 | Missense Mutation | GAG,GGG | E,G 1035 | XP_011522187.1 | |
XM_011523888.2 | 3338 | UTR 3 | XP_011522190.1 | |||
XM_011523889.1 | 3338 | Intron | XP_011522191.1 | |||
XM_011523892.2 | 3338 | Intron | XP_011522194.1 | |||
XM_017024692.1 | 3338 | Intron | XP_016880181.1 | |||
XM_017024693.1 | 3338 | Intron | XP_016880182.1 |
P2RX1 - purinergic receptor P2X 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |