Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGACCTGAAAGTAGTTGTGTAC[A/G]GTAACTGGATTTAGTAGCATTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603616 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RABEP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RABEP1 - rabaptin, RAB GTPase binding effector protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083585.2 | Intron | NP_001077054.1 | ||||
NM_001291581.1 | Intron | NP_001278510.1 | ||||
NM_001291582.1 | Intron | NP_001278511.1 | ||||
NM_004703.5 | Intron | NP_004694.2 | ||||
XM_011524060.1 | Intron | XP_011522362.1 | ||||
XM_017025294.1 | Intron | XP_016880783.1 | ||||
XM_017025295.1 | Intron | XP_016880784.1 |