Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGGCCCCTGGCCCCGGATGC[C/T]TGGTTCTGCCGCTGGTTCCAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603697 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX15B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALOX15B - arachidonate 15-lipoxygenase, type B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039130.1 | 374 | Silent Mutation | GCC,GCT | A,A 86 | NP_001034219.1 | |
NM_001039131.1 | 374 | Silent Mutation | GCC,GCT | A,A 86 | NP_001034220.1 | |
NM_001141.2 | 374 | Silent Mutation | GCC,GCT | A,A 86 | NP_001132.2 |