Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAACCACCTTTTAGAGTGGCGA[C/T]CTCATGGGAGTGGCAGCTTGACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613129 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTC1 - CTS telomere maintenance complex component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025099.5 | 5549 | UTR 3 | NP_079375.3 | |||
XM_006721577.3 | 5549 | UTR 3 | XP_006721640.1 | |||
XM_006721578.3 | 5549 | UTR 3 | XP_006721641.1 | |||
XM_011524010.2 | 5549 | UTR 3 | XP_011522312.1 | |||
XM_011524011.2 | 5549 | UTR 3 | XP_011522313.1 |
LINC00324 - long intergenic non-protein coding RNA 324 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |