Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACGACTACGAGGCTTAGGGCTT[C/T]GTTTTCATCCAATGCCTTTCTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613129 | ||||||||||||||||||||
Literature Links: |
CTC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTC1 - CTS telomere maintenance complex component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025099.5 | 4936 | UTR 3 | NP_079375.3 | |||
XM_006721577.3 | 4936 | UTR 3 | XP_006721640.1 | |||
XM_006721578.3 | 4936 | UTR 3 | XP_006721641.1 | |||
XM_011524010.2 | 4936 | UTR 3 | XP_011522312.1 | |||
XM_011524011.2 | 4936 | UTR 3 | XP_011522313.1 |
LINC00324 - long intergenic non-protein coding RNA 324 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |