Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTCAGCTGTGGGCACAGCTGG[A/G]TGCAGACCACCACTTAGTTGAGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603704 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF222 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF222 - ring finger protein 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146684.2 | 2205 | UTR 3 | NP_001140156.1 | |||
XM_011523978.2 | 2205 | Intron | XP_011522280.1 | |||
XM_011523979.2 | 2205 | Intron | XP_011522281.1 | |||
XM_011523980.2 | 2205 | Intron | XP_011522282.1 | |||
XM_011523981.2 | 2205 | Intron | XP_011522283.1 |
RPL26 - ribosomal protein L26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |