Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTAAACTTTACTAATAAGGTT[A/G]AAGCTTTCTTTTGAACCTCTTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606618 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP14 - dual specificity phosphatase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007026.3 | Intron | NP_008957.1 | ||||
XM_005256977.3 | Intron | XP_005257034.1 | ||||
XM_011524234.1 | Intron | XP_011522536.1 |