Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTTGAAACTGTCTGTTTCTGA[A/G]TTGAATGCCTGCCAGAAATAAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606618 MIM: 607291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP14 - dual specificity phosphatase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNRG - synergin, gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163544.2 | 4029 | Silent Mutation | AAC,AAT | N,N 1196 | NP_001157016.1 | |
NM_001163545.2 | 4029 | Silent Mutation | AAC,AAT | N,N 1195 | NP_001157017.1 | |
NM_001163546.2 | 4029 | Silent Mutation | AAC,AAT | N,N 1139 | NP_001157018.1 | |
NM_001163547.2 | 4029 | Silent Mutation | AAC,AAT | N,N 1068 | NP_001157019.1 | |
NM_007247.5 | 4029 | Silent Mutation | AAC,AAT | N,N 1274 | NP_009178.3 | |
NM_080550.4 | 4029 | Silent Mutation | AAC,AAT | N,N 1184 | NP_542117.3 | |
NM_198882.2 | 4029 | Silent Mutation | AAC,AAT | N,N 1219 | NP_942583.1 | |
XM_005256980.4 | 4029 | Silent Mutation | AAC,AAT | N,N 1375 | XP_005257037.1 | |
XM_017024076.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1398 | XP_016879565.1 | |
XM_017024077.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1397 | XP_016879566.1 | |
XM_017024078.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1386 | XP_016879567.1 | |
XM_017024079.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1377 | XP_016879568.1 | |
XM_017024080.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1371 | XP_016879569.1 | |
XM_017024081.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1370 | XP_016879570.1 | |
XM_017024082.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1363 | XP_016879571.1 | |
XM_017024083.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1353 | XP_016879572.1 | |
XM_017024084.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1352 | XP_016879573.1 | |
XM_017024085.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1342 | XP_016879574.1 | |
XM_017024086.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1330 | XP_016879575.1 | |
XM_017024087.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1329 | XP_016879576.1 | |
XM_017024088.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1320 | XP_016879577.1 | |
XM_017024089.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1318 | XP_016879578.1 | |
XM_017024090.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1297 | XP_016879579.1 | |
XM_017024091.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1296 | XP_016879580.1 | |
XM_017024092.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1296 | XP_016879581.1 | |
XM_017024093.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1262 | XP_016879582.1 | |
XM_017024094.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1261 | XP_016879583.1 | |
XM_017024095.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1253 | XP_016879584.1 | |
XM_017024096.1 | 4029 | Intron | XP_016879585.1 | |||
XM_017024097.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1229 | XP_016879586.1 | |
XM_017024098.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1228 | XP_016879587.1 | |
XM_017024099.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1218 | XP_016879588.1 | |
XM_017024100.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1216 | XP_016879589.1 | |
XM_017024101.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1215 | XP_016879590.1 | |
XM_017024102.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1183 | XP_016879591.1 | |
XM_017024103.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1173 | XP_016879592.1 | |
XM_017024104.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1150 | XP_016879593.1 | |
XM_017024105.1 | 4029 | Intron | XP_016879594.1 | |||
XM_017024106.1 | 4029 | Silent Mutation | AAC,AAT | N,N 1151 | XP_016879595.1 |