Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGGATCACAACATAGACAGTAC[A/G]GCTTTCTTGCTTCCTCTTCGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189907 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNF1B - HNF1 homeobox B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000458.3 | 1948 | UTR 3 | NP_000449.1 | |||
NM_001165923.3 | 1948 | UTR 3 | NP_001159395.1 | |||
NM_001304286.1 | 1948 | UTR 3 | NP_001291215.1 | |||
XM_011525160.1 | 1948 | UTR 3 | XP_011523462.1 | |||
XM_011525161.1 | 1948 | UTR 3 | XP_011523463.1 | |||
XM_011525162.2 | 1948 | Intron | XP_011523464.1 | |||
XM_011525163.2 | 1948 | Intron | XP_011523465.1 | |||
XM_011525164.1 | 1948 | UTR 3 | XP_011523466.1 |