Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGCTGAGGGTGGGGGCCACTTG[A/G]GTGTTTGAGCATTGCCTTTGATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120150 MIM: 600119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL1A1 - collagen type I alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000088.3 | 4811 | UTR 3 | NP_000079.2 | |||
XM_005257058.3 | 4811 | UTR 3 | XP_005257115.2 | |||
XM_005257059.4 | 4811 | UTR 3 | XP_005257116.2 | |||
XM_011524341.1 | 4811 | UTR 3 | XP_011522643.1 |
SGCA - sarcoglycan alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |