Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACCTCTGGGTGGACGCACAGAG[C/T]ACCTGCAGGCGGGAGTGTGAGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WFIKKN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WFIKKN2 - WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175575.5 | 874 | Silent Mutation | AGC,AGT | S,S 60 | NP_783165.1 | |
XM_017024172.1 | 874 | Intron | XP_016879661.1 |