Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACAGGATCTAAGTATTGCCATGG[G/T]GGTGACATCACGCGAAGTCCTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612275 | ||||||||||||||||||||
Literature Links: |
GGNBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GGNBP2 - gametogenetin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024835.4 | 273 | Missense Mutation | GGG,GTG | G,V 73 | NP_079111.1 | |
XM_005257689.2 | 273 | Missense Mutation | GGG,GTG | G,V 73 | XP_005257746.1 | |
XM_011525265.2 | 273 | Missense Mutation | GGG,GTG | G,V 42 | XP_011523567.1 | |
XM_011525266.1 | 273 | Missense Mutation | GGG,GTG | G,V 73 | XP_011523568.1 | |
XM_017025107.1 | 273 | Missense Mutation | GGG,GTG | G,V 42 | XP_016880596.1 |