Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCGTGCGGCGGCGGCGAGCCGGA[A/G]CCGGAACCCGAACCCGAACCCGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PPM1E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPM1E - protein phosphatase, Mg2+/Mn2+ dependent 1E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014906.4 | 225 | Silent Mutation | GAA,GAG | E,E 32 | NP_055721.3 | |
XM_005257169.4 | 225 | Intron | XP_005257226.1 | |||
XM_011524534.2 | 225 | Intron | XP_011522836.1 | |||
XM_017024359.1 | 225 | Intron | XP_016879848.1 |