Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCAAGAGATAACAAAATAGGA[C/T]TGGGAATCAGAACAACCTGAGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBD1 - tubulin delta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193609.1 | Intron | NP_001180538.1 | ||||
NM_001193610.1 | Intron | NP_001180539.1 | ||||
NM_001193611.1 | Intron | NP_001180540.1 | ||||
NM_001193612.1 | Intron | NP_001180541.1 | ||||
NM_001193613.1 | Intron | NP_001180542.1 | ||||
NM_016261.3 | Intron | NP_057345.2 | ||||
XM_005257425.1 | Intron | XP_005257482.1 | ||||
XM_011524871.2 | Intron | XP_011523173.1 | ||||
XM_017024716.1 | Intron | XP_016880205.1 | ||||
XM_017024717.1 | Intron | XP_016880206.1 | ||||
XM_017024718.1 | Intron | XP_016880207.1 | ||||
XM_017024719.1 | Intron | XP_016880208.1 | ||||
XM_017024720.1 | Intron | XP_016880209.1 |