Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCCAAAGATCACATAGTGGTAGT[A/G]TCTTCTGACTTAATTCAGCAGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608938 MIM: 607344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KB1 - ribosomal protein S6 kinase B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272042.1 | Intron | NP_001258971.1 | ||||
NM_001272043.1 | Intron | NP_001258972.1 | ||||
NM_001272044.1 | Intron | NP_001258973.1 | ||||
NM_001272060.1 | Intron | NP_001258989.1 | ||||
NM_003161.3 | Intron | NP_003152.1 | ||||
XM_011525101.2 | Intron | XP_011523403.1 | ||||
XM_011525102.2 | Intron | XP_011523404.1 | ||||
XM_011525103.2 | Intron | XP_011523405.1 | ||||
XM_017024929.1 | Intron | XP_016880418.1 | ||||
XM_017024930.1 | Intron | XP_016880419.1 | ||||
XM_017024931.1 | Intron | XP_016880420.1 | ||||
XM_017024932.1 | Intron | XP_016880421.1 | ||||
XM_017024933.1 | Intron | XP_016880422.1 |
TUBD1 - tubulin delta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |