Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCTAAGGAGAACAGGGACCCCC[C/T]AAAGCCCATGGGAGAGAGGCACGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCARNA20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCARNA20 - small Cajal body-specific RNA 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP32 - ubiquitin specific peptidase 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032582.3 | Intron | NP_115971.2 | ||||
XM_011525371.1 | Intron | XP_011523673.1 | ||||
XM_011525372.1 | Intron | XP_011523674.1 | ||||
XM_011525373.1 | Intron | XP_011523675.1 | ||||
XM_011525374.1 | Intron | XP_011523676.1 | ||||
XM_011525375.1 | Intron | XP_011523677.1 | ||||
XM_011525376.1 | Intron | XP_011523678.1 | ||||
XM_011525378.1 | Intron | XP_011523680.1 | ||||
XM_011525379.2 | Intron | XP_011523681.1 | ||||
XM_017025233.1 | Intron | XP_016880722.1 | ||||
XM_017025234.1 | Intron | XP_016880723.1 |