Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGATGGGCAAGGGGGTGAAGGG[A/G]ACAAGCCACTAGAGCAGGTGAAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607740 | ||||||||||||||||||||
Literature Links: |
C17orf64 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf64 - chromosome 17 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181707.2 | 125 | Missense Mutation | AAC,GAC | N,D 12 | NP_859058.2 | |
XM_005257033.4 | 125 | Intron | XP_005257090.1 | |||
XM_005257034.4 | 125 | Intron | XP_005257091.1 | |||
XM_005257035.4 | 125 | Intron | XP_005257092.1 | |||
XM_005257036.2 | 125 | Missense Mutation | AAC,GAC | N,D 12 | XP_005257093.1 | |
XM_017024166.1 | 125 | Intron | XP_016879655.1 |
USP32 - ubiquitin specific peptidase 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032582.3 | 125 | Intron | NP_115971.2 | |||
XM_011525371.1 | 125 | Intron | XP_011523673.1 | |||
XM_011525372.1 | 125 | Intron | XP_011523674.1 | |||
XM_011525373.1 | 125 | Intron | XP_011523675.1 | |||
XM_011525374.1 | 125 | Intron | XP_011523676.1 | |||
XM_011525375.1 | 125 | Intron | XP_011523677.1 | |||
XM_011525376.1 | 125 | Intron | XP_011523678.1 | |||
XM_011525378.1 | 125 | Intron | XP_011523680.1 | |||
XM_011525379.2 | 125 | Intron | XP_011523681.1 | |||
XM_017025233.1 | 125 | Intron | XP_016880722.1 | |||
XM_017025234.1 | 125 | Intron | XP_016880723.1 |