Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCAGCTCTGACCCTCGCCCCAG[C/G]GGGAGTCTGTAAACATGCAAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601681 MIM: 601736 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTSJ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FTSJ3 - FtsJ homolog 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMC5 - proteasome 26S subunit, ATPase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199163.1 | 1791 | Intron | NP_001186092.1 | |||
NM_002805.5 | 1791 | Intron | NP_002796.4 |
SMARCD2 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098426.1 | 1791 | UTR 3 | NP_001091896.1 | |||
XM_005257604.2 | 1791 | UTR 3 | XP_005257661.2 | |||
XM_017024967.1 | 1791 | UTR 3 | XP_016880456.1 |