Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGGCGCTTCGGGTCCGGCCCA[C/T]CAGCGTGGCAGAGCTGGAGGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KIF19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF19 - kinesin family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153209.3 | 200 | Missense Mutation | ACC,ATC | T,I 21 | NP_694941.2 | |
XM_005257029.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_005257086.1 | |
XM_005257030.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_005257087.1 | |
XM_017024150.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879639.1 | |
XM_017024151.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879640.1 | |
XM_017024152.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879641.1 | |
XM_017024153.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879642.1 | |
XM_017024154.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879643.1 | |
XM_017024155.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879644.1 | |
XM_017024156.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879645.1 | |
XM_017024157.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879646.1 | |
XM_017024158.1 | 200 | Missense Mutation | ACC,ATC | T,I 21 | XP_016879647.1 |