Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAACATGAGCAATTTCTCATTG[C/T]TTAAAGAAAAACTTGGCACATAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOP2A - topoisomerase (DNA) II alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001067.3 | 5226 | UTR 3 | NP_001058.2 | |||
XM_005257632.1 | 5226 | UTR 3 | XP_005257689.1 | |||
XM_011525165.1 | 5226 | Intron | XP_011523467.1 |