Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTACTGTGTCTACTACTACTTG[A/G]GTCTAGCATATACAGCCATGATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDR2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDR2L - cerebellar degeneration related protein 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL58 - mitochondrial ribosomal protein L58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303265.1 | Intron | NP_001290194.1 | ||||
NM_001545.2 | Intron | NP_001536.1 | ||||
XM_017024574.1 | Intron | XP_016880063.1 |