Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATCATCTCCAGAGGCAAGTTCA[C/G]GTAGATCTGAATTTTGTCACACACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL38 - mitochondrial ribosomal protein L38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM65 - tripartite motif containing 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256124.1 | 1919 | UTR 3 | NP_001243053.1 | |||
NM_173547.3 | 1919 | UTR 3 | NP_775818.2 | |||
XM_006721760.3 | 1919 | UTR 3 | XP_006721823.1 | |||
XM_006721761.3 | 1919 | UTR 3 | XP_006721824.1 | |||
XM_011524499.2 | 1919 | Intron | XP_011522801.1 | |||
XM_011524500.2 | 1919 | Intron | XP_011522802.1 | |||
XM_011524501.2 | 1919 | Intron | XP_011522803.1 | |||
XM_011524502.2 | 1919 | Intron | XP_011522804.1 | |||
XM_011524503.2 | 1919 | Intron | XP_011522805.1 | |||
XM_017024343.1 | 1919 | Intron | XP_016879832.1 | |||
XM_017024344.1 | 1919 | UTR 3 | XP_016879833.1 |