Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGAATGCCCACCCTGGGGAAGA[A/C]AGGAGGAATGTCTACAGTAGGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613768 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF213 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF213 - ring finger protein 213 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256071.2 | Intron | NP_001243000.2 | ||||
NM_020954.3 | Intron | NP_066005.2 | ||||
XM_005257545.4 | Intron | XP_005257602.2 | ||||
XM_005257546.4 | Intron | XP_005257603.2 | ||||
XM_006721995.3 | Intron | XP_006722058.1 | ||||
XM_011525084.2 | Intron | XP_011523386.1 | ||||
XM_011525086.2 | Intron | XP_011523388.1 | ||||
XM_011525087.2 | Intron | XP_011523389.1 | ||||
XM_017024905.1 | Intron | XP_016880394.1 |