Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACGGCTCCTACCCGCACTCAT[A/C]GCGGACAGTGCCTGCAGCGGGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611301 MIM: 607643 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAAP100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAAP100 - Fanconi anemia core complex associated protein 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025161.5 | 3582 | UTR 3 | NP_079437.5 | |||
XM_006722111.2 | 3582 | UTR 3 | XP_006722174.1 | |||
XM_006722112.3 | 3582 | UTR 3 | XP_006722175.1 | |||
XM_017025162.1 | 3582 | UTR 3 | XP_016880651.1 | |||
XM_017025163.1 | 3582 | UTR 3 | XP_016880652.1 |
FSCN2 - fascin actin-bundling protein 2, retinal | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |